(S)-2-(1,10-Phenanthrolin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
Supplier: Ambeed
|
Warning
|
A1512207-100MG
A1512207-250MG
A1512207-1G
76667-386EA
124.13
USD
76667-386
77597-850
77624-814
(S)-2-(1,10-Phenanthrolin-2-yl)-4-phenyl-4,5-dihydrooxazole 98%
(S)-2-(1,10-Phenanthrolin-2-yl)-4-phenyl-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₂₁H₁₅N₃O MW: 325.3633 g/mol |
CAS Number:
133435-88-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...