2-((4-Methoxybenzyl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 98%

Supplier: Ambeed
Warning

A1511768-1G A1511768-250MG A1511768-5G
76667-364EA 209 USD
76667-364 76667-366 76667-368
2-((4-Methoxybenzyl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 98%
2-((4-Methoxybenzyl)oxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

Precaución: for Research Use Only

Formula: C₁₈H₂₃BN₂O₄
MW: 342.2 g/mol
MDL Number: MFCD32900083
CAS Number: 2247982-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR