Propyl 3-bromo-4-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
A1509163-1G
A1509163-100MG
76667-362EA
422.24
USD
76667-362
77624-800
Propyl 3-bromo-4-hydroxybenzoate 98%
Propyl 3-bromo-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₃ MW: 259.0965 g/mol |
CAS Number:
37470-59-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...