6,7-Dihydro-5H-pyrazolo[5,1-b][1,3]oxazin-6-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1508657-100MG
A1508657-250MG
76667-348EA
54
USD
76667-348
76667-350
6,7-Dihydro-5H-pyrazolo[5,1-b][1,3]oxazin-6-amine hydrochloride 97%
6,7-Dihydro-5H-pyrazolo[5,1-b][1,3]oxazin-6-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₀ClN₃O |
MDL Number:
MFCD32659924 CAS Number: 2705222-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...