(2R,3S,4R,5S)-2-(Hydroxymethyl)-5-(p-tolylthio)tetrahydrofuran-3,4-diol 98%

Supplier: Ambeed
Warning

A1508551-1G A1508551-5G
76667-344EA 16.98 USD
76667-344 76667-346
(2R,3S,4R,5S)-2-(Hydroxymethyl)-5-(p-tolylthio)tetrahydrofuran-3,4-diol 98%
(2R,3S,4R,5S)-2-(Hydroxymethyl)-5-(p-tolylthio)tetrahydrofuran-3,4-diol

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄S
MW: 256.31 g/mol
CAS Number: 1131610-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR