rel-tert-Butyl ((3aS,6aR)-2-benzylhexahydrocyclopenta[c]pyrrol-3a(1H)-yl)carbamate 97%

Supplier: Ambeed
Warning

A1508505-100MG A1508505-250MG
76667-340EA 51.99 USD
76667-340 76667-342
rel-tert-Butyl ((3aS,6aR)-2-benzylhexahydrocyclopenta[c]pyrrol-3a(1H)-yl)carbamate 97%
rel-tert-Butyl ((3aS,6aR)-2-benzylhexahydrocyclopenta[c]pyrrol-3a(1H)-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₉H₂₈N₂O₂
MW: 316.4378 g/mol
CAS Number: 2828438-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR