rel-2-(tert-Butyl) 7a-methyl (3aR,7aS)-5-methylhexahydro-1H-pyrrolo[3,4-c]pyridine-2,7a-dicarboxylate 97%

Supplier: Ambeed
Warning

A1508208-100MG A1508208-250MG
76667-332EA 51.99 USD
76667-332 76667-334
rel-2-(tert-Butyl) 7a-methyl (3aR,7aS)-5-methylhexahydro-1H-pyrrolo[3,4-c]pyridine-2,7a-dicarboxylate 97%
rel-2-(tert-Butyl) 7a-methyl (3aR,7aS)-5-methylhexahydro-1H-pyrrolo[3,4-c]pyridine-2,7a-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₆N₂O₄
MW: 298.3779 g/mol
CAS Number: 2757083-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR