4-(10,15,20-Tri-p-tolylporphyrin-5-yl)phenol 95%

Supplier: Ambeed
Warning

A1508184-100MG A1508184-1G A1508184-250MG A1508184-50MG
76667-324EA 258.86 USD
76667-324 76667-326 76667-328 76667-330
4-(10,15,20-Tri-p-tolylporphyrin-5-yl)phenol 95%
4-(10,15,20-Tri-p-tolylporphyrin-5-yl)phenol

Precaución: for Research Use Only

Formula: C₄₇H₃₆N₄O
MW: 672.8150 g/mol
MDL Number: MFCD32643948
CAS Number: 57412-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR