4-(10,15,20-Tris(4-bromophenyl)porphyrin-5-yl)phenol 95%

Supplier: Ambeed
Warning

A1508182-100MG A1508182-1G A1508182-250MG A1508182-50MG
76667-308EA 259.93 USD
76667-308 76667-310 76667-312 76667-314
4-(10,15,20-Tris(4-bromophenyl)porphyrin-5-yl)phenol 95%
4-(10,15,20-Tris(4-bromophenyl)porphyrin-5-yl)phenol

Precaución: for Research Use Only

Formula: C₄₄H₂₇Br₃N₄O
MW: 867.4234 g/mol
CAS Number: 147951-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR