5,10,15,20-Tetrakis(4-ethylphenyl)porphyrin 95%

Supplier: Ambeed
Warning

A1508181-100MG A1508181-1G A1508181-250MG
76667-302EA 56.23 USD
76667-302 76667-304 76667-306
5,10,15,20-Tetrakis(4-ethylphenyl)porphyrin 95%
5,10,15,20-Tetrakis(4-ethylphenyl)porphyrin

Precaución: for Research Use Only

Formula: C₅₂H₄₆N₄
MW: 726.94 g/mol
CAS Number: 146943-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR