(S)-2-(2,6-Dioxopiperidin-3-yl)-4-hydroxyisoindoline-1,3-dione 95%

Supplier: Ambeed
Warning

A1508166-100MG A1508166-1G A1508166-250MG
76667-296EA 183 USD
76667-296 76667-298 76667-300
(S)-2-(2,6-Dioxopiperidin-3-yl)-4-hydroxyisoindoline-1,3-dione 95%
(S)-2-(2,6-Dioxopiperidin-3-yl)-4-hydroxyisoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂O₅
CAS Number: 2357105-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR