Methyl 1-tosyl-1,2,3,6-tetrahydropyridine-4-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A1507728-100MG
A1507728-250MG
76667-278EA
51.99
USD
76667-278
76667-280
Methyl 1-tosyl-1,2,3,6-tetrahydropyridine-4-carboxylate 98%
Methyl 1-tosyl-1,2,3,6-tetrahydropyridine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₁₇NO₄S MW: 295.35 g/mol |
CAS Number:
1615213-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...