3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridine-4-carboxamide 95%

Supplier: Ambeed
Warning

A1507603-100MG A1507603-250MG
76667-274EA 51.99 USD
76667-274 76667-276
3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridine-4-carboxamide 95%
3-Isopropyl-6,7-dihydro-5H-cyclopenta[c]pyridine-4-carboxamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O
MW: 204.26 g/mol
CAS Number: 2757730-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR