Iridium-(2,2'-bipyridine-κN1,κN1')bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507445-100MG A1507445-1G A1507445-250MG
76667-262EA 186.72 USD
76667-262 76667-264 76667-266
Iridium-(2,2'-bipyridine-κN1,κN1')bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate 95%
Iridium-(2,2'-bipyridine-κN1,κN1')bis[3,5-difluoro-2-(5-fluoro-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₂H₁₈F₁₂IrN₄P
MW: 909.6837 g/mol
CAS Number: 2517366-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR