Iridium-(1,10-phenanthroline-κN1,κN10)bis[5-methyl-2-(4-methyl-2-pyridinyl-κN)phenyl-κC]-(OC-6-33)-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507444-100MG A1507444-1G A1507444-250MG A1507444-50MG
76667-254EA 204.76 USD
76667-254 76667-256 76667-258 76667-260
Iridium-(1,10-phenanthroline-κN1,κN10)bis[5-methyl-2-(4-methyl-2-pyridinyl-κN)phenyl-κC]-(OC-6-33)-hexafluorophosphate 95%
Iridium-(1,10-phenanthroline-κN1,κN10)bis[5-methyl-2-(4-methyl-2-pyridinyl-κN)phenyl-κC]-(OC-6-33)-hexafluorophosphate

Precaución: for Research Use Only

CAS Number: 2828447-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR