Iridium-(2,2'-bipyridine-κN1,κN1')bis[5-(1,1-dimethylethyl)-2-[4-(1,1-dimethylethyl)-2-pyridinyl-κN]phenyl-κC]-(OC-6-33)-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507441-100MG A1507441-1G A1507441-250MG
76667-242EA 85 USD
76667-242 76667-244 76667-246
Iridium-(2,2'-bipyridine-κN1,κN1')bis[5-(1,1-dimethylethyl)-2-[4-(1,1-dimethylethyl)-2-pyridinyl-κN]phenyl-κC]-(OC-6-33)-hexafluorophosphate 95%
Iridium-(2,2'-bipyridine-κN1,κN1')bis[5-(1,1-dimethylethyl)-2-[4-(1,1-dimethylethyl)-2-pyridinyl-κN]phenyl-κC]-(OC-6-33)-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₄₈H₅₆F₆IrN₄P
MW: 1026.1662 g/mol
CAS Number: 808142-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR