Iridium-(2,2'-bipyridine-κN1,κN1')bis[(3-fluoro-5-trifluoromethyl)-2-(5-trifluoromethyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507439-100MG A1507439-1G A1507439-250MG
76667-228EA 194 USD
76667-228 76667-230 76667-232
Iridium-(2,2'-bipyridine-κN1,κN1')bis[(3-fluoro-5-trifluoromethyl)-2-(5-trifluoromethyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate 95%
Iridium-(2,2'-bipyridine-κN1,κN1')bis[(3-fluoro-5-trifluoromethyl)-2-(5-trifluoromethyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate

Precaución: for Research Use Only

CAS Number: 2828437-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR