Iridium-(1,10-phenanthroline-κN1,κN10)bis[3,5-difluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507438-100MG A1507438-1G A1507438-250MG A1507438-50MG
76667-220EA 93 USD
76667-220 76667-222 76667-224 76667-226
Iridium-(1,10-phenanthroline-κN1,κN10)bis[3,5-difluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate 95%
Iridium-(1,10-phenanthroline-κN1,κN10)bis[3,5-difluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-hexafluorophosphate

Precaución: for Research Use Only

CAS Number: 2828437-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR