Iridium-(2,2'-bipyridine-κN1,κN1')bis[3,5-difluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-(OC-6-33)-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507437-100MG A1507437-1G A1507437-250MG
76667-214EA 197 USD
76667-214 76667-216 76667-218
Iridium-(2,2'-bipyridine-κN1,κN1')bis[3,5-difluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-(OC-6-33)-hexafluorophosphate 95%
Iridium-(2,2'-bipyridine-κN1,κN1')bis[3,5-difluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC]-(OC-6-33)-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₂₄F₁₀IrN₄P
MW: 901.7560 g/mol
CAS Number: 1335047-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR