Iridium-bis[5-fluoro-2-(2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507436-100MG A1507436-1G A1507436-250MG
76667-208EA 196.27 USD
76667-208 76667-210 76667-212
Iridium-bis[5-fluoro-2-(2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate 95%
Iridium-bis[5-fluoro-2-(2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₂₂F₈IrN₄P
MW: 861.7433 g/mol
CAS Number: 1426547-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR