Iridium-bis[5-fluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507435-100MG A1507435-1G A1507435-250MG A1507435-50MG
76667-200EA 227 USD
76667-200 76667-202 76667-204 76667-206
Iridium-bis[5-fluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate 95%
Iridium-bis[5-fluoro-2-(5-methyl-2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₆H₂₆F₈IrN₄P
CAS Number: 855523-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR