Iridium-bis[3,5-difluoro-2-(2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate 95%

Supplier: Ambeed
Warning

A1507433-100MG A1507433-1G A1507433-250MG A1507433-50MG
76667-192EA 92 USD
76667-192 76667-194 76667-196 76667-198
Iridium-bis[3,5-difluoro-2-(2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate 95%
Iridium-bis[3,5-difluoro-2-(2-pyridinyl-κN)phenyl-κC](1,10-phenanthroline-κN1,κN10)-(OC-6-13)-hexafluorophosphate

Precaución: for Research Use Only

Formula: C₃₄H₂₀F₁₀IrN₄P
CAS Number: 1426548-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR