(3aR,6R,6aR)-6-(((tert-Butyldimethylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol 98%

Supplier: Ambeed
Warning

A1506052-5G A1506052-1G
76667-178EA 206 USD
76667-178 77597-812
(3aR,6R,6aR)-6-(((tert-Butyldimethylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol 98%
(3aR,6R,6aR)-6-(((tert-Butyldimethylsilyl)oxy)methyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol

Precaución: for Research Use Only

Formula: C₁₄H₂₈O₅Si
MW: 304.4546 g/mol
MDL Number: MFCD01075716
CAS Number: 217309-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR