9,9-Dimethyl-N2,N7-diphenyl-9H-fluorene-2,7-diamine 95%

Supplier: Ambeed
Warning

A1506037-100MG A1506037-1G A1506037-250MG
76667-172EA 10 USD
76667-172 76667-174 76667-176
9,9-Dimethyl-N2,N7-diphenyl-9H-fluorene-2,7-diamine 95%
9,9-Dimethyl-N2,N7-diphenyl-9H-fluorene-2,7-diamine

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂
CAS Number: 354987-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR