2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-(dibenzo[b,d]furan-3-yl)-1,3,5-triazine 95%

Supplier: Ambeed
Warning

A1505896-1G A1505896-250MG
76667-160EA 124.13 USD
76667-160 76667-162
2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-(dibenzo[b,d]furan-3-yl)-1,3,5-triazine 95%
2-([1,1'-Biphenyl]-3-yl)-4-chloro-6-(dibenzo[b,d]furan-3-yl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₇H₁₆ClN₃O
MW: 433.8884 g/mol
CAS Number: 2226747-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR