2,6-Difluoro-4-(trans-4-propylcyclohexyl)benzonitrile 95%

Supplier: Ambeed
Warning

A1505727-25G A1505727-5G A1505727-100G
76667-142EA 26 USD
76667-142 76667-144 77875-875
2,6-Difluoro-4-(trans-4-propylcyclohexyl)benzonitrile 95%
2,6-Difluoro-4-(trans-4-propylcyclohexyl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₉F₂N
MDL Number: MFCD11053470
CAS Number: 167306-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR