2,2'-Dihydroxybiphenyl 99.5+% (by GC)

Supplier: Ambeed
Warning

Synonyms: 2,2'-Biphenol, 2,2-Biphenol, Biphenyl-2,2'-diol

A1505289-100G A1505289-25G A1505289-500G
76667-132EA 25 USD
76667-132 76667-134 76667-136
2,2'-Dihydroxybiphenyl 99.5+% (by GC)
2,2'-Dihydroxybiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MDL Number: MFCD00002210
CAS Number: 1806-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR