6-Chloro-2-propyl-9H-purine 95%

Supplier: Ambeed
Warning

A1505251-100MG A1505251-250MG
76667-128EA 53.05 USD
76667-128 76667-130
6-Chloro-2-propyl-9H-purine 95%
6-Chloro-2-propyl-9H-purine

Precaución: for Research Use Only

Formula: C₈H₉ClN₄
MW: 196.63 g/mol
CAS Number: 757184-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR