7-Amino-6-bromo-2,3-dihydro-1H-indene-4-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A1504622-100MG
A1504622-250MG
76667-106EA
54.11
USD
76667-106
76667-108
7-Amino-6-bromo-2,3-dihydro-1H-indene-4-carbonitrile 95%
7-Amino-6-bromo-2,3-dihydro-1H-indene-4-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₀H₉BrN₂ MW: 237.0959 g/mol |
CAS Number:
2757083-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...