3-Bromo-9H-pyrido[2,3-b]indole 98%

Supplier: Ambeed
Warning

A1504472-1G A1504472-250MG
76667-102EA 146.41 USD
76667-102 76667-104
3-Bromo-9H-pyrido[2,3-b]indole 98%
3-Bromo-9H-pyrido[2,3-b]indole

Precaución: for Research Use Only

Formula: C₁₁H₇BrN₂
MW: 247.0907 g/mol
MDL Number: MFCD31922754
CAS Number: 1428799-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR