16-Iodo-2,5,8,11,14-pentaoxahexadecane 98+% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilized with Copper chip

A1504293-1G A1504293-250MG A1504293-5G
76667-094EA 110 USD
76667-094 76667-096 77875-849
16-Iodo-2,5,8,11,14-pentaoxahexadecane 98+% stabilized
16-Iodo-2,5,8,11,14-pentaoxahexadecane

Precaución: for Research Use Only

Formula: C₁₁H₂₃IO₅
MDL Number: MFCD31656944
CAS Number: 104518-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR