1-Iodo-2-(2-(2-methoxyethoxy)ethoxy)ethane ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilized with Copper chip

A1504291-1G A1504291-5G
76667-092EA 94.43 USD
76667-092 77567-158
1-Iodo-2-(2-(2-methoxyethoxy)ethoxy)ethane ≥98% stabilized
1-Iodo-2-(2-(2-methoxyethoxy)ethoxy)ethane

Precaución: For research use only.

Formula: C₇H₁₅IO₃
MW: 274.09 g/mol
MDL Number: MFCD26911322
CAS Number: 62999-96-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR