(R)-1-(3-Nitro-5-(trifluoromethyl)phenyl)ethan-1-amine hydrochloride 98%

Supplier: Ambeed
Warning

A1504190-1G A1504190-250MG
76667-082EA 560.16 USD
76667-082 76667-084
(R)-1-(3-Nitro-5-(trifluoromethyl)phenyl)ethan-1-amine hydrochloride 98%
(R)-1-(3-Nitro-5-(trifluoromethyl)phenyl)ethan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀ClF₃N₂O₂
MW: 270.64 g/mol
MDL Number: MFCD32265411
CAS Number: 2230840-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR