(2-Fluoro-5-methoxy-4-methylphenyl)boronic acid 95%

Supplier: Ambeed
Warning

A1503934-1G A1503934-250MG A1503934-5G
76667-074EA 218 USD
76667-074 76667-076 76667-078
(2-Fluoro-5-methoxy-4-methylphenyl)boronic acid 95%
(2-Fluoro-5-methoxy-4-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD19440300
CAS Number: 900175-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR