1-((Benzyloxy)methyl)pyrimidine-2,4(1H,3H)-dione 95%

Supplier: Ambeed
Warning

A1503751-100MG A1503751-1G A1503751-250MG
76667-066EA 103.97 USD
76667-066 76667-068 76667-070
1-((Benzyloxy)methyl)pyrimidine-2,4(1H,3H)-dione 95%
1-((Benzyloxy)methyl)pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₃
MW: 232.2353 g/mol
CAS Number: 80140-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR