(2S,2'S)-2,2'-Bipyrrolidine dihydrochloride 98+%

Supplier: Ambeed
Warning

A1503664-100MG A1503664-1G A1503664-250MG
76667-060EA 71.09 USD
76667-060 76667-062 76667-064
(2S,2'S)-2,2'-Bipyrrolidine dihydrochloride 98+%
(2S,2'S)-2,2'-Bipyrrolidine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂
MW: 213.1479 g/mol
CAS Number: 2757287-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR