5,7-Di-tert-butyl-3-(3,4-dichlorophenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%

Supplier: Ambeed
Danger

A1503654-250MG A1503654-100MG
76667-058EA 46 USD
76667-058 77597-788
5,7-Di-tert-butyl-3-(3,4-dichlorophenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%
5,7-Di-tert-butyl-3-(3,4-dichlorophenyl)benzo[d]oxazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₁H₂₄BCl₂F₄NO
CAS Number: 2750161-94-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR