5,7-Di-tert-butyl-3-(4-(trifluoromethyl)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%
Supplier: Ambeed
|
Danger
|
A1503653-1G
A1503653-250MG
A1503653-5G
A1503653-100MG
76667-054EA
200.52
USD
76667-054
76667-056
77549-884
77597-786
5,7-Di-tert-butyl-3-(4-(trifluoromethyl)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate 97%
5,7-Di-tert-butyl-3-(4-(trifluoromethyl)phenyl)benzo[d]oxazol-3-ium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₂₂H₂₅BF₇NO MW: 463.23 g/mol |
MDL Number:
MFCD34596159 CAS Number: 2750161-92-1 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...