N4,N4-Bis(4'-amino-[1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine 97%

Supplier: Ambeed
Warning

A1503160-100MG A1503160-1G A1503160-250MG
76667-030EA 26.53 USD
76667-030 76667-032 76667-034
N4,N4-Bis(4'-amino-[1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine 97%
N4,N4-Bis(4'-amino-[1,1'-biphenyl]-4-yl)-[1,1'-biphenyl]-4,4'-diamine

Precaución: for Research Use Only

Formula: C₃₆H₃₀N₄
MW: 518.65 g/mol
CAS Number: 2607105-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR