4'',4''''',4'''''''',4'''''''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1':4',1''-terphenyl]-4-carbaldehyde)) 97%

Supplier: Ambeed
Warning

A1503159-100MG A1503159-1G A1503159-250MG
76667-024EA 119.89 USD
76667-024 76667-026 76667-028
4'',4''''',4'''''''',4'''''''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1':4',1''-terphenyl]-4-carbaldehyde)) 97%
4'',4''''',4'''''''',4'''''''''''-(Ethene-1,1,2,2-tetrayl)tetrakis(([1,1':4',1''-terphenyl]-4-carbaldehyde))

Precaución: for Research Use Only

Formula: C₇₈H₅₂O₄
MW: 1053.2451 g/mol
CAS Number: 2567920-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR