1-((2,3-Difluoro-4-(trans-4-pentylcyclohexyl)phenyl)ethynyl)-4-ethoxy-2,3-difluorobenzene 95%

Supplier: Ambeed
Warning

A1502049-1G A1502049-25G
76667-000EA 8 USD
76667-000 77875-798
1-((2,3-Difluoro-4-(trans-4-pentylcyclohexyl)phenyl)ethynyl)-4-ethoxy-2,3-difluorobenzene 95%
1-((2,3-Difluoro-4-(trans-4-pentylcyclohexyl)phenyl)ethynyl)-4-ethoxy-2,3-difluorobenzene

Precaución: for Research Use Only

Formula: C₂₇H₃₀F₄O
MW: 446.5201 g/mol
CAS Number: 928208-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR