2,3-Difluoro-4-propoxy-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl 95%
Supplier: Ambeed
|
Warning
|
A1502018-25G
A1502018-5G
76666-992EA
41.38
USD
76666-992
76666-994
2,3-Difluoro-4-propoxy-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl 95%
2,3-Difluoro-4-propoxy-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₂₄H₃₀F₂O MW: 372.4912 g/mol |
CAS Number:
442912-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...