rel-(3aR,6aR)-Hexahydrocyclopenta[c]pyrrole-3a(1H)-carboxylic acid 98%

Supplier: Ambeed
Warning

A1501892-100MG A1501892-250MG A1501892-50MG
76666-964EA 177.18 USD
76666-964 76666-966 76666-968
rel-(3aR,6aR)-Hexahydrocyclopenta[c]pyrrole-3a(1H)-carboxylic acid 98%
rel-(3aR,6aR)-Hexahydrocyclopenta[c]pyrrole-3a(1H)-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MW: 155.19 g/mol
CAS Number: 1212067-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR