Rel-4-butoxy-2,3-difluoro-4'-((1s,4r)-4-propylcyclohexyl)-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A1501851-25G A1501851-5G
76666-956EA 41.38 USD
76666-956 76666-958
Rel-4-butoxy-2,3-difluoro-4'-((1s,4r)-4-propylcyclohexyl)-1,1'-biphenyl 98%
Rel-4-butoxy-2,3-difluoro-4'-((1s,4r)-4-propylcyclohexyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₂₅H₃₂F₂O
MW: 386.5178 g/mol
CAS Number: 323178-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR