Rel-(1's,4'r)-4-(2,3-difluoro-4-propoxyphenyl)-4'-propyl-1,1'-bi(cyclohexane) 98%

Supplier: Ambeed
Warning

A1501848-5G A1501848-100G A1501848-25G
76666-954EA 24 USD
76666-954 77875-759 77875-760
Rel-(1's,4'r)-4-(2,3-difluoro-4-propoxyphenyl)-4'-propyl-1,1'-bi(cyclohexane) 98%
Rel-(1's,4'r)-4-(2,3-difluoro-4-propoxyphenyl)-4'-propyl-1,1'-bi(cyclohexane)

Precaución: for Research Use Only

Formula: C₂₄H₃₆F₂O
MW: 378.53 g/mol
CAS Number: 473257-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR