N-(4-(Phenylamino)pent-3-en-2-ylidene)aniline hydrochloride 95%

Supplier: Ambeed
Warning

A1501766-1G A1501766-250MG A1501766-5G
76666-950EA 43 USD
76666-950 77875-749 77875-750
N-(4-(Phenylamino)pent-3-en-2-ylidene)aniline hydrochloride 95%
N-(4-(Phenylamino)pent-3-en-2-ylidene)aniline hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClN₂
CAS Number: 19164-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR