3-Bromo-5-chloro-2-iodoaniline ≥96%

Supplier: Ambeed
Warning

A1501547-100MG A1501547-1G A1501547-250MG
76666-944EA 21.22 USD
76666-944 76666-946 76666-948
3-Bromo-5-chloro-2-iodoaniline ≥96%
3-Bromo-5-chloro-2-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClIN
MW: 332.3641 g/mol
MDL Number: MFCD28250212
CAS Number: 1823408-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR