trans-1-Cinnamylpiperazine 95%
Supplier: Ambeed
Synonyms:
(E)-1-Cinnamylpiperazine
A1501282-1G
A1501282-25G
76666-936EA
10
USD
76666-936
76666-938
trans-1-Cinnamylpiperazine 95%
trans-1-Cinnamylpiperazine
Precaución: for Research Use Only
Formula:
C₁₃H₁₈N₂ MW: 202.30 g/mol |
MDL Number:
MFCD00063251 CAS Number: 87179-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...