N1,N3-Bis(4-(tert-butyl)phenyl)-5-methylbenzene-1,3-diamine 98%

Supplier: Ambeed
Warning

A1501189-1G A1501189-5G
76666-932EA 13 USD
76666-932 77875-733
N1,N3-Bis(4-(tert-butyl)phenyl)-5-methylbenzene-1,3-diamine 98%
N1,N3-Bis(4-(tert-butyl)phenyl)-5-methylbenzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₂
MW: 386.5723 g/mol
CAS Number: 2757731-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR