(4S,4'S)-4,4'-Diisopropyl-1,1'-di-m-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole 97%
Supplier: Ambeed
|
Warning
|
A1501171-1G
A1501171-250MG
A1501171-100MG
76666-926EA
745
USD
76666-926
76666-928
77875-725
(4S,4'S)-4,4'-Diisopropyl-1,1'-di-m-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole 97%
(4S,4'S)-4,4'-Diisopropyl-1,1'-di-m-tolyl-4,4',5,5'-tetrahydro-1H,1'H-2,2'-biimidazole
Precaución: for Research Use Only
Formula:
C₂₆H₃₄N₄ |
CAS Number:
2374958-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...